Learn More
Sino Biological Human CD177 MAb 25TS (Sino Biological)
Supplier:  Sino Biological 14501MM05A25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD177 / PRV-1 / PRV1 (rh CD177 / PRV-1 / PRV1; Catalog#14501-H08H; AAH29167.1; Met1-Gly407) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-237-2086