Learn More
Sino Biological Recombinant Anti-CD177 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 14501R021100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD177 / PRV-1 / PRV1 (rh CD177 / PRV-1 / PRV1; Catalog#14501-H08H; AAH29167.1; Met1-Gly407).
Catalog No. 50-255-4130