Learn More
Sino Biological Anti-CD177 Antibody (PE), Mouse Monoclonal
Supplier:  Sino Biological 14501MM05P25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD177 / PRV-1 / PRV1 (rh CD177 / PRV-1 / PRV1; Catalog#14501-H08H; AAH29167.1; Met1-Gly407) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-255-4132