Learn More
Sino Biological Anti-CD177 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 14501T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CD177 / PRV-1 / PRV1 (rh CD177 / PRV-1 / PRV1; Catalog#14501-H08H; AAH29167.1; Met1-Gly407). CD177 / PRV-1 / PRV1 specific IgG was purified by Human CD177 / PRV-1 / PRV1 affinity chromatography.
Catalog No. 50-255-4128