Learn More
ACROBiosystems Human CD19 2.5mg
Catalog No. 502534474
Encompass_Preferred
ActiveMax Human CD19 uBeads premium grade (for cells) are uniform superparamagnetic beads of 5.5 µm with streptavidin on its surface and coupled with biotinylated Human CD19 Protein expressed from human 293 cells (HEK293) and contains AA Pro 20 - Lys 291 (Accession # P15391-1).
Catalog No. 50-253-4474 Supplier ACROBiosystems Supplier No. MBSC0022.5MG
ActiveMax Human CD19 uBeads premium grade (for cells) are uniform superparamagnetic beads of 5.5 µm with streptavidin on its surface and coupled with biotinylated Human CD19 Protein expressed from human 293 cells (HEK293) and contains AA Pro 20 - Lys 291 (Accession # P15391-1).