Learn More
ACROBiosystems Alexa Fluor 647-Labeled Human CD19 (20-291) Protein, His TagStar Staining
Supplier:  ACROBiosystems CD9HA2H625UG
Encompass_Preferred
Alexa Fluor 647-Labeled Human CD19 (20-291), His Tag (CD9-HA2H6) is produced via conjugation of AF647 to Human CD19 (20-291), His Tag with a new generation site-specific technology under Star Staining labeling platform. Human CD19 (20-291), His Tag is expressed from human 293 cells (HEK293). It contains AA Pro 20 - Lys 291 (Accession # P15391-1).
Catalog No. 50-233-0966