Learn More
ACROBiosystems Human CD19-coupled Magnetic Beads, 2 mg, Superparamagnetic, Biotinylated CD19 to Streptavidin
Supplier:  ACROBiosystems MBSK0052MG
Encompass_Preferred
The Human CD19-coupled Magnetic Beads (2 mg) provide an efficient solution for isolating anti-CD19 antibodies from biological samples. These superparamagnetic beads are designed for optimal binding and purification through a simple workflow, suitable for high-throughput applications. The product features a uniform size and specialized surface coating for reduced non-specific binding, enhancing assay accuracy.
  • Specific antibody capture using biotinylated Human CD19 protein conjugated to streptavidin magnetic beads.
  • High reproducibility ensured through uniform bead size and narrow distribution.
  • Reduced non-specific binding with optimized surface chemistry for enhanced specificity.
  • Streamlined workflow with a rapid "bind-wash-elute" process, eliminating the need for sophisticated equipment.
  • High-throughput compatibility; ready-to-use format saves time and effort.
Catalog No. NC2093151