Learn More
Sino Biological Recombinant Anti-CD19 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 11880R10920
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD19 (rh CD19; Catalog#11880-H08H; P15391-1; Met1-Lys291).
Catalog No. 50-241-1719