Learn More
Sino Biological Human CD19 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 11880MM17100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD19 (rh CD19; Catalog#11880-H08H; P15391-1; Met1-Lys291). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-2749