Learn More
Sino Biological Anti-CD19 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11880T56100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CD19 (rh CD19; Catalog#11880-H08H; P15391-1; Met1-Lys291). CD19 specific IgG was purified by Human CD19 affinity chromatography.
Catalog No. 50-255-4494