Learn More
Sino Biological Recombinant Human CD19 Protein (ECD, His & AVI Tag), Biotinylated
Supplier:  Sino Biological 11880H49HB20
Encompass_Preferred
A DNA sequence encoding the Human CD19 (P15391-1) (Met1-Lys291) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-237-4094