Learn More
BIOHIPPO Human CD1A Microsample
SDP
Catalog No. 5000314966
Encompass
Human CD1A Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CD1A in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 063 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CD1A Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CD1A Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CD1A biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14966 Supplier BIOHIPPO Supplier No. BHE15212372
Human CD1A Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CD1A in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 063 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CD1A Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CD1A Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CD1A biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color