Learn More
Sino Biological Recombinant Human CD2 Protein (ECD, hFc Tag), HPLC-verified
Supplier:  Sino Biological 10982H02H200
Encompass_Preferred
A DNA sequence encoding the human CD2 (NP_001758.2) (Met1-Asp209) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-255-4996