Learn More
ACROBiosystems ACROBiosystems Human CD2 / SRBC Protein, His Tag
Catalog No. 502106545
Encompass_Preferred
Recombinant Protein;50UG;Human CD2, His Tag (CD2-H5226) is expressed from human 293 cells (HEK293). It contains AA Lys 25 - Asp 209 (Accession # AAH33583).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 22.3 kDa. The protein migrates as 33-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD58, Fc Tag (Cat. No. LF3-H5256) at 5 μg/mL (100 μL/well) can bind Human CD2, His Tag (Cat. No. CD2-H5226) with a linear range of 0.02-0.313 μg/mL (QC tested).
Catalog No. 50-210-6545 Supplier ACROBiosystems Supplier No. CD2H522650UG
Recombinant Protein;50UG;Human CD2, His Tag (CD2-H5226) is expressed from human 293 cells (HEK293). It contains AA Lys 25 - Asp 209 (Accession # AAH33583).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 22.3 kDa. The protein migrates as 33-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD58, Fc Tag (Cat. No. LF3-H5256) at 5 μg/mL (100 μL/well) can bind Human CD2, His Tag (Cat. No. CD2-H5226) with a linear range of 0.02-0.313 μg/mL (QC tested).