Learn More
ACROBiosystems Human CD20 20ug
Catalog No. 502535396
Encompass_Preferred
Human CD20 Full Length Protein FlagHis Tag (CD0-H52D5) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Pro 297 (Accession # P11836-1).
Catalog No. 50-253-5396 Supplier ACROBiosystems Supplier No. CD0H52D520UG
Human CD20 Full Length Protein FlagHis Tag (CD0-H52D5) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Pro 297 (Accession # P11836-1).