Learn More
ACROBiosystems ACROBiosystems Human CD20 Full Length Protein-VLP (HEK293)
Supplier:  ACROBiosystems CDPH52P6500UG
Encompass_Preferred
Recombinant Protein;500UG;Human CD20 Full Length Protein-VLP (CDP-H52P6) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Pro 297 (Accession # M4ZHZ6-1 ).;This protein carries no "tag". The protein has a calculated MW of 33 kDa.
Catalog No. 50-210-6953