Learn More
Sino Biological Anti-CD200 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10886MM17100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD200 (rh CD200; Catalog#10886-H08H; NP_005935.4; Met1-Gly232). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-5107