Learn More
Sino Biological Recombinant Anti-CD200 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10886R005100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD200 (rh CD200; Catalog#10886-H08H; NP_005935.4; Met 1-Gly 232).
Catalog No. 50-255-5104