Learn More
Sino Biological Recombinant Human CD200 Protein (ECD, hFc Tag), HPLC-verified
Supplier:  Sino Biological 10886H02H1
Encompass_Preferred
A DNA sequence encoding the human CD200 (XP_003825138.1) (Met1-Gly232) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-222-3576