Learn More
Sino Biological Anti-CD200 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10886T32100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CD200 (rh CD200; Catalog#10886-H08H; NP_005935.4; Met1-Gly232). CD200 specific IgG was purified by Human CD200 affinity chromatography.
Catalog No. 50-255-5100