Learn More
Sino Biological Recombinant Human CD200 Protein (ECD, hFc & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 10886H41HB100
Encompass_Preferred
A DNA sequence encoding the human CD200 ( NP_005935.4) (Met1-Gly232) was expressed with a C-terminal Fc region of human IgG1 tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-5109