Learn More
ACROBiosystems Human CD200 R1 / CRTR2 Protein, His Tag
Catalog No. 502054754
Encompass_Preferred
Recombinant Protein;1MG;Human CD200 R1, His Tag (CR2-H52H6) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Leu 266 (Accession # AAQ19772.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 28.7 kDa. The protein migrates as 48-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD200, Fc Tag (Cat. No. OX2-H5251) at 5 μg/mL (100 μL/well) can bind Human CD200 R1, His Tag (Cat. No. CR2-H52H6) with a linear range of 20-78 ng/mL (QC tested).
Catalog No. 50-205-4754 Supplier ACROBiosystems Supplier No. CR2H52H61MG
Recombinant Protein;1MG;Human CD200 R1, His Tag (CR2-H52H6) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Leu 266 (Accession # AAQ19772.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 28.7 kDa. The protein migrates as 48-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD200, Fc Tag (Cat. No. OX2-H5251) at 5 μg/mL (100 μL/well) can bind Human CD200 R1, His Tag (Cat. No. CR2-H52H6) with a linear range of 20-78 ng/mL (QC tested).