Learn More
Sino Biological Anti-CD200R Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11218RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CD200R1 extracellular domain (rh CD200R1; Catalog#11218-H08H; AAI43394.1; Met 1-Leu 266). CD200R1 specific IgG was purified by human CD200R1 affinity chromatography.
Catalog No. 50-255-4781