Learn More
Sino Biological Recombinant Anti-DC-SIGN/CD209 Antibody (HRP), Rabbit Monoclonal
Supplier:  Sino Biological 10200R059H100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD209 / DC-SIGN, (rh CD209 / DC-SIGN; Catalog#10200-H01H; NP_066978.1; Lys62-Ala404), and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-241-1160