Learn More
Sino Biological Recombinant Human Siglec-2/CD22 Protein (ECD, hFc & AVI Tag), Biotinylated
Supplier:  Sino Biological 11958H41HB100
Encompass_Preferred
A DNA sequence encoding the human CD22 (NP_001172029.1) (Met1-Arg687) was expressed with a C-terminal Fc region of human IgG1 tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-4483