Learn More
Sino Biological Recombinant Human Siglec-2/CD22 Protein (aa 176-687, His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 11958H08H1B20
Encompass_Preferred
A DNA sequence encoding the human CD22 (Q32M46) (Trp176-Arg687) was fused with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-255-4484