Learn More
American Research Products Inc Human CD226 antigen CD226 EL
Catalog No. 5000327432
Encompass
Human CD226 antigen (CD226) ELISA kit from CUSABIO Detection Range 0 9 ng/mL-60 ng/mL Sensitivity 0 22 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27432 Supplier American Research Products Inc Supplier No. CSBEL004901HU
Human CD226 antigen (CD226) ELISA kit from CUSABIO Detection Range 0 9 ng/mL-60 ng/mL Sensitivity 0 22 ng/mL Sample serum plasma tissue homogenates Method Sandwich