Learn More
Sino Biological Recombinant Human DNAM-1/CD226 Protein (ECD, hFc Tag)
Supplier:  Sino Biological 10565H02H100
Encompass_Preferred
A DNA sequence encoding the human CD226 (NP_006557.2) (Met1-Asn247) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-255-5261