Learn More
Sino Biological Recombinant Human DNAM-1/CD226 Protein (ECD, His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 10565H08HB100
Encompass_Preferred
A DNA sequence encoding the human CD226 (NP_006557.2) (Met1-Asn247) was expressed with a C-terminal polyhistidine tag. The purified protein was biotinylated in vitro.
Catalog No. 50-255-5260