Learn More
ACROBiosystems ACROBiosystems Human CD229 / SLAMF3 Protein, His Tag
Catalog No. 502106577
Encompass_Preferred
Recombinant Protein;1MG;Human CD229, His Tag (CD9-H52H6) is expressed from human 293 cells (HEK293). It contains AA Lys 48 - Lys 454 (Accession # Q9HBG7-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 46.7 kDa. The protein migrates as 60-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD229, His Tag (Cat. No. CD9-H52H6) at 5 μg/mL (100 μL/well) can bind Biotinylated Human CD229, His,Avitag (Cat. No. CD9-H82E6) with a linear range of 0.039-2.5 μg/mL (QC tested).
Catalog No. 50-210-6577 Supplier ACROBiosystems Supplier No. CD9H52H61MG
Recombinant Protein;1MG;Human CD229, His Tag (CD9-H52H6) is expressed from human 293 cells (HEK293). It contains AA Lys 48 - Lys 454 (Accession # Q9HBG7-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 46.7 kDa. The protein migrates as 60-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD229, His Tag (Cat. No. CD9-H52H6) at 5 μg/mL (100 μL/well) can bind Biotinylated Human CD229, His,Avitag (Cat. No. CD9-H82E6) with a linear range of 0.039-2.5 μg/mL (QC tested).