Learn More
ACROBiosystems ACROBiosystems Human CD23 / Fc epsilon RII Protein, His Tag
Supplier:  ACROBiosystems CD3H5249100UG
Encompass_Preferred
Recombinant Protein;100UG;Human CD23, His Tag (CD3-H5249) is expressed from human 293 cells (HEK293). It contains AA Asp 48 - Ser 321 (Accession # P06734-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 32.9 kDa. The protein migrates as 40-44 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6547