Learn More
Sino Biological Human CD244 Protein 20ug (Sino Biological)
Supplier:  Sino Biological 10042H41HB20
Encompass_Preferred
A DNA sequence encoding the human CD244 (NP_057466.1) (Met1-Arg221) was expressed with a c-terminal AVI tagged Fc region of human IgG1 at the C-terminus (Fc-AVI). The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-237-3883