Learn More
Sino Biological Human CD244 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 10042H49HB100
Encompass_Preferred
A DNA sequence encoding the human CD244 (NP_057466.1) (Met1-Arg221) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-237-2048