Learn More
Sino Biological Human CD247 MAb 25TS (Sino Biological)
Supplier:  Sino Biological 13142MM06P25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD3 zeta / CD247 (rh CD3 zeta / CD247; Catalog#13142-H09E; P20963-1; Arg52-Arg164) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-237-1869