Learn More
BIOHIPPO Human CD247 Microsample
SDP
Catalog No. 5000313873
Encompass
Human CD247 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CD247 in Serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 06 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CD247 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CD247 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CD247 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13873 Supplier BIOHIPPO Supplier No. BHE15210492
Human CD247 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CD247 in Serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 06 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CD247 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CD247 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CD247 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color