Learn More
Sino Biological Human CD247 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 13142T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CD247/CD3-ZETA (rh CD247/CD3-ZETA; Catalog#13142-H09E; P20963-1; Arg52-Arg164). CD247/CD3-ZETA specific IgG was purified by Human CD247/CD3-ZETA affinity chromatography.
Catalog No. 50-237-2571