Learn More
Sino Biological Anti-CD3 zeta/CD247 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 200144T46100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human CD3 zeta/CD247 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-255-3972