Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human CD248 Synthetic Peptide

Catalog No. PEP0553 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP0553 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP0553 Supplier Invitrogen™ Supplier No. PEP0553
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic Peptide

This peptide corresponds to 14 amino acids near the carboxy terminus of human TEM1. PEP-0553 can be used as a blocking peptide with polyclonal antibody PA5-20436.

CD248 (CD248 Molecule) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include calcium ion binding and extracellular matrix binding. An important paralog of this gene is CD93. CD248 may play a role in tumor angiogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 57124
Species Human
Purification Method purified
Accession Number Q9HCU0
Common Name CD248
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias 2610111G01Rik; AI842296; CD164 sialomucin-like 1; CD164L1; Cd248; CD248 antigen, endosialin; CD248 molecule; CD248 molecule, endosialin; endosialin; MGC119478; MGC119479; TEM1; tumor endothelial marker 1
Gene Symbol CD248
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.