Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human BAFF (BLyS) Recombinant Protein

Catalog No. 5018544
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-185-44 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-185-44 Supplier Invitrogen™ Supplier No. 14894380
Only null left
Add to Cart
Add to Cart

Recombinant Protein

Description: Recombinant human BAFF (B-cell activating factor), also referred to as BlyS, is a type II transmembrane protein of the TNF superfamily. BAFF is found on T cells, macrophages and dendritic cells. BAFF contains a cytoplasmic domain, transmembrane domain and extracellular domain which can be cleaved between amino acid 133 and 134 to produce a soluble form detectable in serum. Receptors for BAFF include TACI, BAMBI and BAFFR. BAFF/BLyS is a B lymphocyte stimulatory molecule; it induces B cell proliferation and immunoglobulin secretion. In addition BAFF can provide a co-stimulation signal to T lymphocytes. Elevated levels of BAFF has been implicated in the pathogenesis of some autoimmune diseases (particularly) B cell diseases such as systemic lupus erythaematosus. Human and mouse BAFF share 86% amino acid sequence identity. Applications Reported: Recombinant human BAFF is biologically active. Applications Tested: The ED50, as determined by a mouse splenocytes survival assay, is 1.6 μg/mL. Source: E. coli expressed amino acids Ala134-Leu285 accession # Q9Y275. Bioactivity: The ED50, as determined by a mouse splenocyte survival assay, is 1.6 μg/mL. Endotoxin: Less than 0.40 ng/ug as determined by the LAL assay. Purity: >90% as determined by reducing SDS-PAGE. Molecular Weight: 18.5 kDa. Storage and handling: For best recovery, quick-spin vial prior to opening. Use in a sterile environment. Purity: Greater than 90%, as determined by SDS-PAGE.

BAFF is a membrane bound cytokine expressed by B cell lineage cells. The soluble form of the cytokine binds to the receptors TNFRSF13B/TACI, TNFRSF17/BCMA, and TNFRSF13C/BAFFR. BAFF is a potent B cell activator, and important for the proliferation and differentiation of B cells. BAFF is also expressed by T lymphocytes and has been shown to promote T cell activation and survival. Isoforms representing alternatively spliced, and glycosylated variants have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9Y275
Concentration 0.1 mg/mL
For Use With (Application) Bioactivity, Functional Assay
Formulation PBS with no preservative
Gene ID (Entrez) 10673
Molecular Weight (g/mol) 18.5 kDa
Name Human BAFF (BLyS)
Purification Method Purified
Quantity 25 μg
Source E. coli
Storage Requirements -80°C
Regulatory Status RUO
Endotoxin Concentration <0.40 ng/μg
Gene Alias ApoL related ligand TALL-1; b cell-activating factor; B lymphocyte stimulator; BAFF; B-cell-activating factor; B-lymphocyte stimulator; BLYS; CD257; D8Ertd387e; delta BAFF; Delta4 BAFF; dendritic cell-derived TNF-like molecule; DTL; RGD1561519; TALL1; TALL-1; THANK; TNF- and APOL-related leukocyte expressed ligand 1; TNF and ApoL-related leukocyte expressed ligand 1; TNF homolog that activates apoptosis; Tnfsf13b; TNFSF20; TNLG7A; tumor necrosis factor (ligand) superfamily member 13b; tumor necrosis factor (ligand) superfamily, member 13b; tumor necrosis factor (ligand) superfamily, member 20; tumor necrosis factor ligand 7A; tumor necrosis factor ligand superfamily member 13B; Tumor necrosis factor ligand superfamily member 13b, membrane form; Tumor necrosis factor ligand superfamily member 13b, soluble form; tumor necrosis factor superfamily member 13b; tumor necrosis factor-like protein ZTNF4; UNQ401/PRO738; ZTNF4
Common Name BAFF (BLyS)
Gene Symbol TNFSF13B
Biological Activity ED50 = 1.6 μg/mL; determined by a mouse splenocyte survival assay.
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Sequence Human BAFF, amino acids Ala134-Leu285 (Accession # Q9Y275)
Content And Storage -80°C
Activity ED50 = 1.6 μg/mL; determined by a mouse splenocyte survival assay.
Expression System E. coli
Form Liquid
Protein Subtype Recombinant
Purity or Quality Grade >90% by reducing SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.