Learn More
Sino Biological Recombinant Human CD27/TNFRSF7 Protein (ECD, hFc Tag)
Supplier:  Sino Biological 10039H02H100
Encompass_Preferred
A DNA sequence encoding the human CD27 (NP_001233.1) (Met1-Ile192) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-255-5526