Learn More
Sino Biological Recombinant Human CD27/TNFRSF7 Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 10039H08B1B20
Encompass_Preferred
A DNA sequence encoding the human CD27(P26842) (Met1-Ile192) was expressed with a C-terminal polyhistidine tag. The purified protein was biotinylated in vitro.
Catalog No. 50-255-5524