Learn More
Sino Biological Human CD274 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 10084RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human B7-H1 (rh B7-H1; Catalog#10084-H08H; NP_054862.1; Met 1-Thr 239). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-237-1454