Learn More
Sino Biological Anti-PD-L1/B7-H1 Antibody (Biotin), Rabbit Polyclonal
Supplier:  Sino Biological 10084RP02B20
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human B7-H1 / CD274 / PD-L1 (rh B7-H1; Catalog#10084-H08H; NP_054862.1; Met 1-Thr 239). B7-H1 specific IgG was purified by Human B7-H1 affinity chromatography and then biotinylated.
Catalog No. 50-255-5490