Learn More
Sino Biological Recombinant Human PD-L1/B7-H1 Protein (AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 10084H17HB100
Encompass_Preferred
A DNA sequence encoding the Human CD274 (NP_054862.1) (Met1-Thr239) was fused with an AVI tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-5496