Learn More
Sino Biological Recombinant Human PD-L1/B7-H1 Protein (ECD, hFc Tag), HPLC-verified
Supplier:  Sino Biological 10084H02H1
Encompass_Preferred
A DNA sequence encoding the N-terminal segment (Met 1-Thr 239) of the extracellular domain of human B7-H1 (NP_054862.1) was expressed with C-terminal fused Fc region of human IgG1.
Catalog No. 50-255-5499