Learn More
Sino Biological Anti-B7-H3 Antibody (FITC), Mouse Monoclonal
Supplier:  Sino Biological 11188MM06F100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human B7-H3 / CD276 (rh B7-H3 / CD276; Catalog#11188-H08H; Q5ZPR3-1; Met 1-Thr 461) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-241-1801