Learn More
Sino Biological Anti-B7-H3 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 11188MM01100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human B7-H3 / CD276 (rh B7-H3 / CD276; Catalog#11188-H08H; Q5ZPR3-1; Met1-Thr461). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-1757