Learn More
Sino Biological Recombinant Human B7-H3 Protein (ECD, His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 11188H27HB100
Encompass_Preferred
A DNA sequence encoding the human CD276 (Q5ZPR3) (Met1-Thr461) was expressed with a c-terminal polyhistidine tagged AVI tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-4805