Learn More
Sino Biological Recombinant Human B7-H3 Protein (ECD, hFc Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 11188H02HB20
Encompass_Preferred
A DNA sequence encoding the human CD276 (NP_001019907.1) (Met1-Thr461) was expressed with the Fc region of human IgG1 at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-255-4806