Learn More
BIOHIPPO Human CD27BP Microsample
SDP
Catalog No. 5000315666
Encompass
Human CD27BP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CD27BP in Tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 071 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CD27BP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CD27BP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CD27BP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15666 Supplier BIOHIPPO Supplier No. BHE15210942
Human CD27BP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CD27BP in Tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 071 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CD27BP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CD27BP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CD27BP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color